Product Details

SNP ID
rs77353614
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:204194779 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTTTACAGAAAGTGTTCCCAGGAC[A/G]GTGCTAAGGCGCTGAAGCAAGGGCA
Phenotype
MIM: 603286
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GOLT1A PubMed Links

Gene Details

Gene
GOLT1A
Gene Name
golgi transport 1A
There are no transcripts associated with this gene.

Gene
KISS1
Gene Name
KiSS-1 metastasis-suppressor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002256.3 Intron NP_002247.3

View Full Product Details