Product Details

SNP ID
rs74731791
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:154167548 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATTACTATCCTGAGTAACCTGTA[C/T]TAAATCCATCACTCTGGTAGAATCA
Phenotype
MIM: 191030
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TPM3 PubMed Links

Gene Details

Gene
TPM3
Gene Name
tropomyosin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001043351.1 1364 Intron NP_001036816.1
NM_001043352.1 1364 Intron NP_001036817.1
NM_001043353.1 1364 Intron NP_001036818.1
NM_001278188.1 1364 Intron NP_001265117.1
NM_001278189.1 1364 Intron NP_001265118.1
NM_001278190.1 1364 Intron NP_001265119.1
NM_001278191.1 1364 Intron NP_001265120.1
NM_152263.3 1364 UTR 3 NP_689476.2
NM_153649.3 1364 Intron NP_705935.1

View Full Product Details