Product Details
- SNP ID
-
rs77832800
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
16
- Location
-
Chr.1:155085361 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGTTCCAGCGCTACAGCGCCTTCTC[G/T]CTGGGCTACGAGTTCCACGCCGGCC
- Phenotype
-
MIM: 601381
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
EFNA3
PubMed Links
Gene Details
- Gene
- EFNA3
- Gene Name
- ephrin A3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004952.4 |
469 |
Silent Mutation |
TCG,TCT |
S133S |
NP_004943.1 |
View Full Product Details