Product Details

SNP ID
rs77292424
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:156073039 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATCGCCTCAGTGCTCTTGAAGCT[C/G]TGAAGGTGAGAAGCGTGTGTGCATG
Phenotype
MIM: 150330 MIM: 611007 MIM: 612942
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LMNA PubMed Links

Gene Details

Gene
LMNA
Gene Name
lamin A/C
There are no transcripts associated with this gene.

Gene
MEX3A
Gene Name
mex-3 RNA binding family member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001093725.1 5098 UTR 3 NP_001087194.1
Gene
RAB25
Gene Name
RAB25, member RAS oncogene family
There are no transcripts associated with this gene.

View Full Product Details