Product Details

SNP ID
rs78256534
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:156368187 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTGGAATTGAGAAGCCCTCCTTG[C/T]ATTCAGCCTCCCTCCGCTCCAGCTG
Phenotype
MIM: 607079
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RHBG PubMed Links

Gene Details

Gene
RHBG
Gene Name
Rh family B glycoprotein (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256395.1 Intron NP_001243324.1
NM_001256396.1 Intron NP_001243325.1
NM_020407.4 Intron NP_065140.3
XM_011509799.2 Intron XP_011508101.1
XM_011509800.2 Intron XP_011508102.1
XM_017001857.1 Intron XP_016857346.1
XM_017001858.1 Intron XP_016857347.1
XM_017001859.1 Intron XP_016857348.1

View Full Product Details