Product Details

SNP ID
rs77839882
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:16064540 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGTCCACTTTGAAGCATTCAGCC[A/G]GGATCTCATATGCTGCGATGTGTCT
Phenotype
MIM: 602023
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLCNKB PubMed Links

Gene Details

Gene
CLCNKB
Gene Name
chloride voltage-gated channel Kb
There are no transcripts associated with this gene.

Gene
FAM131C
Gene Name
family with sequence similarity 131 member C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182623.2 Intron NP_872429.2

View Full Product Details