Product Details

SNP ID
rs77071615
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:18840027 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGTGGGACTGAGGCCCGTGGCCA[A/G]CATGCCAATTACCACAATGACCATT
Phenotype
MIM: 606226
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TAS1R2 PubMed Links

Gene Details

Gene
TAS1R2
Gene Name
taste 1 receptor member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152232.2 2093 Silent Mutation CTG,TTG L698L NP_689418.2

View Full Product Details