Product Details

SNP ID
rs75510165
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:21598818 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCCTGATATCTGGGAGGCAAGGC[A/G]GGCTGTGAAGAGACCATTGCAATTC
Phenotype
MIM: 600278
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RAP1GAP PubMed Links

Gene Details

Gene
RAP1GAP
Gene Name
RAP1 GTPase activating protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145657.1 Intron NP_001139129.1
NM_001145658.1 Intron NP_001139130.1
NM_002885.2 Intron NP_002876.2
XM_005245955.3 Intron XP_005246012.2
XM_006710804.2 Intron XP_006710867.2
XM_006710805.2 Intron XP_006710868.2
XM_017001965.1 Intron XP_016857454.1
XM_017001966.1 Intron XP_016857455.1
XM_017001967.1 Intron XP_016857456.1
XM_017001968.1 Intron XP_016857457.1
XM_017001969.1 Intron XP_016857458.1
XM_017001970.1 Intron XP_016857459.1
XM_017001971.1 Intron XP_016857460.1
XM_017001972.1 Intron XP_016857461.1
XM_017001973.1 Intron XP_016857462.1
XM_017001974.1 Intron XP_016857463.1
XM_017001975.1 Intron XP_016857464.1
XM_017001976.1 Intron XP_016857465.1
XM_017001977.1 Intron XP_016857466.1
XM_017001978.1 Intron XP_016857467.1
XM_017001979.1 Intron XP_016857468.1
XM_017001980.1 Intron XP_016857469.1
XM_017001981.1 Intron XP_016857470.1
XM_017001982.1 Intron XP_016857471.1
XM_017001983.1 Intron XP_016857472.1
XM_017001984.1 Intron XP_016857473.1
XM_017001985.1 Intron XP_016857474.1
XM_017001986.1 Intron XP_016857475.1
XM_017001987.1 Intron XP_016857476.1
XM_017001988.1 Intron XP_016857477.1
XM_017001989.1 Intron XP_016857478.1
XM_017001990.1 Intron XP_016857479.1
XM_017001991.1 Intron XP_016857480.1

View Full Product Details