Product Details
- SNP ID
-
rs79171291
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
2
- Location
-
Chr.1:21690023 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATGACGCCAAGGGTGCCTAGGGTG[G/T]CACAGTCATCACTTAAAATCTTTCC
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
USP48
PubMed Links
Gene Details
- Gene
- USP48
- Gene Name
- ubiquitin specific peptidase 48
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001032730.1 |
3758 |
Intron |
|
|
NP_001027902.1 |
XM_005246009.2 |
3758 |
Missense Mutation |
GAC,GCC |
D935A |
XP_005246066.1 |
XM_006710951.2 |
3758 |
Missense Mutation |
GAC,GCC |
D985A |
XP_006711014.2 |
XM_006710955.3 |
3758 |
Missense Mutation |
GAC,GCC |
D921A |
XP_006711018.1 |
XM_011542262.2 |
3758 |
Missense Mutation |
GAC,GCC |
D986A |
XP_011540564.1 |
XM_011542263.2 |
3758 |
Missense Mutation |
GAC,GCC |
D986A |
XP_011540565.1 |
XM_011542264.1 |
3758 |
Missense Mutation |
GAC,GCC |
D920A |
XP_011540566.1 |
XM_011542265.2 |
3758 |
Missense Mutation |
GAC,GCC |
D860A |
XP_011540567.1 |
XM_011542267.2 |
3758 |
Missense Mutation |
GAC,GCC |
D573A |
XP_011540569.1 |
XM_011542268.2 |
3758 |
Missense Mutation |
GAC,GCC |
D573A |
XP_011540570.1 |
XM_017002495.1 |
3758 |
Missense Mutation |
GAC,GCC |
D920A |
XP_016857984.1 |
XM_017002496.1 |
3758 |
Intron |
|
|
XP_016857985.1 |
XM_017002497.1 |
3758 |
Missense Mutation |
GAC,GCC |
D573A |
XP_016857986.1 |
View Full Product Details