Product Details

SNP ID
rs79545952
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:238139969 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTGACGCGAAACCCTCCAGAACG[C/T]TGCCTTTGGCTGGCCGTTTCTGTTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ESPNL PubMed Links

Gene Details

Gene
ESPNL
Gene Name
espin-like
There are no transcripts associated with this gene.

Gene
KLHL30
Gene Name
kelch like family member 30
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198582.3 Intron NP_940984.3

View Full Product Details