Product Details

SNP ID
rs76298340
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:238169210 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACACCTGGTTTGCTATAGGAAGA[C/T]AGTTACCAAATGAATGGTTCTATTT
Phenotype
MIM: 615099
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ERFE PubMed Links

Gene Details

Gene
ERFE
Gene Name
erythroferrone
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291832.1 Intron NP_001278761.1
Gene
ILKAP
Gene Name
ILK associated serine/threonine phosphatase
There are no transcripts associated with this gene.

View Full Product Details