Product Details

SNP ID
rs74728218
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:8013759 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCAATGATTGTAATGATTGGCAG[A/G]GTGGTATCCAGGGACCCTGGCCCAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC00299 PubMed Links

Gene Details

Gene
LINC00299
Gene Name
long intergenic non-protein coding RNA 299
There are no transcripts associated with this gene.

View Full Product Details