Product Details

SNP ID
rs78528665
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:8856936 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGGTAGGGTTAGCCTTATCAGC[C/T]GGAAAACACTTACTTTTAGATAAAT
Phenotype
MIM: 611949
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MBOAT2 PubMed Links

Gene Details

Gene
MBOAT2
Gene Name
membrane bound O-acyltransferase domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321265.1 3519 UTR 3 NP_001308194.1
NM_001321266.1 3519 UTR 3 NP_001308195.1
NM_001321267.1 3519 UTR 3 NP_001308196.1
NM_138799.3 3519 UTR 3 NP_620154.2
XM_011510313.1 3519 Intron XP_011508615.1
XM_011510314.2 3519 Intron XP_011508616.1

View Full Product Details