Product Details

SNP ID
rs77558140
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:5694915 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTTGTTCCCTTATACCTCCTTGA[C/T]TGAATACCAGACAGCCTAGACCTCA
Phenotype
MIM: 600898
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC01248 PubMed Links

Gene Details

Gene
LINC01248
Gene Name
long intergenic non-protein coding RNA 1248
There are no transcripts associated with this gene.

Gene
SOX11
Gene Name
SRY-box 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003108.3 2249 UTR 3 NP_003099.1

View Full Product Details