Product Details

SNP ID
rs79772157
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:14636283 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTTCACTGCTTTGGTGGAAATTG[A/G]TGGAAATTGCTAGCAGGTTCCACGA
Phenotype
MIM: 611234
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM84A PubMed Links

Gene Details

Gene
FAM84A
Gene Name
family with sequence similarity 84 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145175.2 2592 UTR 3 NP_660158.2
Gene
LOC653602
Gene Name
uncharacterized LOC653602
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291410.1 2592 Intron NP_001278339.1

View Full Product Details