Product Details

SNP ID
rs77425662
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:159717329 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGCCATATTTTGTAATTTACCATA[G/T]GTTCTTTAGACCACTATAAGAATAG
Phenotype
MIM: 605683 MIM: 613334
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
BAZ2B PubMed Links

Gene Details

Gene
BAZ2B
Gene Name
bromodomain adjacent to zinc finger domain 2B
There are no transcripts associated with this gene.

Gene
MARCH7
Gene Name
membrane associated ring-CH-type finger 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282805.1 Intron NP_001269734.1
NM_001282806.1 Intron NP_001269735.1
NM_001282807.1 Intron NP_001269736.1
NM_022826.3 Intron NP_073737.1
XM_005246773.2 Intron XP_005246830.1
XM_005246774.1 Intron XP_005246831.1
XM_017004718.1 Intron XP_016860207.1
XM_017004719.1 Intron XP_016860208.1
XM_017004720.1 Intron XP_016860209.1
XM_017004721.1 Intron XP_016860210.1

View Full Product Details