Product Details

SNP ID
rs74850973
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:165481287 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTCTTCCTGTAGTGTGAGGTAGA[A/C]TCTTGGCAAGATACGGTCACTTAAT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CSRNP3 PubMed Links

Gene Details

Gene
CSRNP3
Gene Name
cysteine and serine rich nuclear protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172173.1 Intron NP_001165644.1
NM_024969.3 Intron NP_079245.2
XM_005246865.4 Intron XP_005246922.1

View Full Product Details