Product Details
- SNP ID
-
rs79165518
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:174348983 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTTTTTCTTTCTTTTCTTCCTTTG[G/T]ATTTTTTTTTCTTTATTGTTACTCT
- Phenotype
-
MIM: 605228
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CIR1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs73975114] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CIR1
- Gene Name
- corepressor interacting with RBPJ, 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004882.3 |
|
Intron |
|
|
NP_004873.3 |
View Full Product Details