Product Details

SNP ID
rs76616433
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:174436377 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCAATTAGCCAGCTCACCAGGTC[A/C]CAGCCACAGAAAGTTCCAGCAGAAG
Phenotype
MIM: 614967
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GPR155 PubMed Links

Gene Details

Gene
GPR155
Gene Name
G protein-coupled receptor 155
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033045.3 2790 Missense Mutation TGG,TGT W784C NP_001028217.1
NM_001267050.1 2790 Missense Mutation TGG,TGT W784C NP_001253979.1
NM_001267051.1 2790 Missense Mutation TGG,TGT W756C NP_001253980.1
NM_152529.6 2790 Missense Mutation TGG,TGT W784C NP_689742.4
XM_017003485.1 2790 Missense Mutation TGG,TGT W784C XP_016858974.1
XM_017003486.1 2790 Missense Mutation TGG,TGT W784C XP_016858975.1
XM_017003487.1 2790 Missense Mutation TGG,TGT W784C XP_016858976.1
Gene
SCRN3
Gene Name
secernin 3
There are no transcripts associated with this gene.

View Full Product Details