Product Details

SNP ID
rs80306085
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:151836956 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTAAATACTTTAATGCGCATAAA[C/T]AAGAAACCAGTAAAGCATATAGAAA
Phenotype
MIM: 608960 MIM: 601949
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARL5A PubMed Links

Gene Details

Gene
ARL5A
Gene Name
ADP ribosylation factor like GTPase 5A
There are no transcripts associated with this gene.

Gene
CACNB4
Gene Name
calcium voltage-gated channel auxiliary subunit beta 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011511795.2 4514 UTR 3 XP_011510097.1
XM_011511796.2 4514 UTR 3 XP_011510098.1
XM_011511797.2 4514 UTR 3 XP_011510099.1
XM_011511798.2 4514 Intron XP_011510100.1
XM_017004880.1 4514 UTR 3 XP_016860369.1
XM_017004881.1 4514 UTR 3 XP_016860370.1
XM_017004882.1 4514 UTR 3 XP_016860371.1
XM_017004883.1 4514 UTR 3 XP_016860372.1
XM_017004884.1 4514 UTR 3 XP_016860373.1
XM_017004885.1 4514 UTR 3 XP_016860374.1
XM_017004886.1 4514 UTR 3 XP_016860375.1
XM_017004887.1 4514 Intron XP_016860376.1
XM_017004888.1 4514 Intron XP_016860377.1

View Full Product Details