Product Details

SNP ID
rs79637495
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:179948192 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAATCAAGTTGAAGGAACTCCTAA[G/T]TGCCAAAGCTAAGATGATTTGAGCA
Phenotype
MIM: 615186
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CWC22 PubMed Links

Gene Details

Gene
CWC22
Gene Name
CWC22 homolog, spliceosome-associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020943.2 Intron NP_065994.1
XM_005246726.2 Intron XP_005246783.1

View Full Product Details