Product Details

SNP ID
rs76917094
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:189666348 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTGGAAGATGAAAGAGGCAATGT[G/T]TTTCTATGGAATGGAGAAATTTTTA
Phenotype
MIM: 609803
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ANKAR PubMed Links

Gene Details

Gene
ANKAR
Gene Name
ankyrin and armadillo repeat containing
There are no transcripts associated with this gene.

Gene
ASNSD1
Gene Name
asparagine synthetase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019048.2 1166 Silent Mutation GTG,GTT V72V NP_061921.1
XM_017004381.1 1166 Silent Mutation GTG,GTT V72V XP_016859870.1
XM_017004382.1 1166 Silent Mutation GTG,GTT V72V XP_016859871.1

View Full Product Details