Product Details

SNP ID
rs79064785
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:201458702 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTCTCTTTTCCCCACTGTAGTCA[C/T]TAGACCTTTTTGTATCTACATACCA
Phenotype
MIM: 607333 MIM: 607334
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
STRADB PubMed Links

Gene Details

Gene
STRADB
Gene Name
STE20-related kinase adaptor beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206864.1 Intron NP_001193793.1
NM_018571.5 Intron NP_061041.2
XM_005246669.1 Intron XP_005246726.1
XM_017004438.1 Intron XP_016859927.1
Gene
TRAK2
Gene Name
trafficking kinesin protein 2
There are no transcripts associated with this gene.

View Full Product Details