Product Details

SNP ID
rs79677156
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:208128339 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTGGGCAGCTCGTAGAGGACCCAG[A/C]AGCCCTCCAGCACGTGGAGGGAACG
Phenotype
MIM: 123680 MIM: 123690
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CRYGC PubMed Links

Gene Details

Gene
CRYGC
Gene Name
crystallin gamma C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020989.3 427 Missense Mutation NP_066269.1
XM_011510662.1 427 Missense Mutation XP_011508964.1
Gene
CRYGD
Gene Name
crystallin gamma D
There are no transcripts associated with this gene.

Gene
LOC100507443
Gene Name
uncharacterized LOC100507443
There are no transcripts associated with this gene.

View Full Product Details