Product Details

SNP ID
rs79824607
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:216674656 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGATATCTTCAGCCTTAAAGAA[C/T]AGTCACAAGGCCACCCATTGGTTGA
Phenotype
MIM: 146734
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IGFBP5 PubMed Links

Gene Details

Gene
IGFBP5
Gene Name
insulin like growth factor binding protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000599.3 3688 UTR 3 NP_000590.1

View Full Product Details