Product Details

SNP ID
rs79691056
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:218577243 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTATTTTTAAAAAATTGTATTC[A/T]TCCCATGACAGGTGGGTATTCAGTT
Phenotype
MIM: 612054
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CNOT9 PubMed Links

Gene Details

Gene
CNOT9
Gene Name
CCR4-NOT transcription complex subunit 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271634.1 Intron NP_001258563.1
NM_001271635.1 Intron NP_001258564.1
NM_005444.2 Intron NP_005435.1
XM_011512138.2 Intron XP_011510440.1
XM_017005248.1 Intron XP_016860737.1
XM_017005249.1 Intron XP_016860738.1
Gene
USP37
Gene Name
ubiquitin specific peptidase 37
There are no transcripts associated with this gene.

View Full Product Details