Product Details

SNP ID
rs77827981
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:222572000 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCCCTGGCAAAGATCTCTGCACA[G/T]CGCCCGGGGAAGAAAGCAGGCCCTG
Phenotype
MIM: 609690 MIM: 612827
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FARSB PubMed Links

Gene Details

Gene
FARSB
Gene Name
phenylalanyl-tRNA synthetase beta subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005687.4 1923 Silent Mutation CGA,CGC R547R NP_005678.3
XM_006712169.2 1923 Silent Mutation CGA,CGC R448R XP_006712232.1
XM_011510466.2 1923 Silent Mutation CGA,CGC R448R XP_011508768.1
XM_017003110.1 1923 Silent Mutation CGA,CGC R448R XP_016858599.1
Gene
SGPP2
Gene Name
sphingosine-1-phosphate phosphatase 2
There are no transcripts associated with this gene.

View Full Product Details