Product Details

SNP ID
rs79783536
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:226736155 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCATCGTACCATCTACTGATGAG[A/G]AAGATATGAGGTCCTAGTTGTGAAT
Phenotype
MIM: 147545
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IRS1 PubMed Links

Gene Details

Gene
IRS1
Gene Name
insulin receptor substrate 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005544.2 3898 UTR 3 NP_005535.1

View Full Product Details