Product Details

SNP ID
rs77065513
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:230999775 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTAGATCAATGGAACGTGGCAGC[C/T]CCAATTCCAGGAATGTCAGTGCAGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SPATA3 PubMed Links

Gene Details

Gene
SPATA3
Gene Name
spermatogenesis associated 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_139073.3 Intron NP_620712.2
XM_005246303.2 Intron XP_005246360.1
XM_017003363.1 Intron XP_016858852.1
Gene
SPATA3-AS1
Gene Name
SPATA3 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details