Product Details

SNP ID
rs77853740
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:231204609 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTACTGTGTGCCAGGCACTGGTC[C/T]AGGCACAGGGGAACACCTGTCAGGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARMC9 PubMed Links

Gene Details

Gene
ARMC9
Gene Name
armadillo repeat containing 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271466.2 Intron NP_001258395.1
NM_001291656.1 Intron NP_001278585.1
NM_025139.5 Intron NP_079415.3
XM_011511905.1 Intron XP_011510207.1
XM_011511906.1 Intron XP_011510208.1
XM_011511907.1 Intron XP_011510209.1
XM_011511908.2 Intron XP_011510210.1
XM_011511909.2 Intron XP_011510211.1
XM_011511910.1 Intron XP_011510212.1
XM_011511911.1 Intron XP_011510213.1
XM_011511912.1 Intron XP_011510214.1
XM_011511913.2 Intron XP_011510215.1
XM_011511914.2 Intron XP_011510216.1
XM_011511915.2 Intron XP_011510217.1
XM_011511916.2 Intron XP_011510218.1
XM_011511919.2 Intron XP_011510221.1
XM_017005018.1 Intron XP_016860507.1
XM_017005019.1 Intron XP_016860508.1
XM_017005020.1 Intron XP_016860509.1
XM_017005021.1 Intron XP_016860510.1
XM_017005022.1 Intron XP_016860511.1
XM_017005023.1 Intron XP_016860512.1
XM_017005024.1 Intron XP_016860513.1
XM_017005025.1 Intron XP_016860514.1
XM_017005026.1 Intron XP_016860515.1

View Full Product Details