Product Details

SNP ID
rs75201778
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:231975348 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTATTATATATTATATCTCTTTGA[C/G]GCTAAGAATCCAGGGAAGGGAAAGC
Phenotype
MIM: 614184
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DIS3L2 PubMed Links

Gene Details

Gene
DIS3L2
Gene Name
DIS3 like 3'-5' exoribonuclease 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001257281.1 Intron NP_001244210.1
NM_001257282.1 Intron NP_001244211.1
NM_152383.4 Intron NP_689596.4

View Full Product Details