Product Details

SNP ID
rs74913005
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:232554510 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGTAGCTCATACACACCATAGAT[A/G]CTTGGTAGATGTTGGTAATTTTTAA
Phenotype
MIM: 100730 MIM: 605895 MIM: 612972
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHRNG PubMed Links

Gene Details

Gene
CHRNG
Gene Name
cholinergic receptor nicotinic gamma subunit
There are no transcripts associated with this gene.

Gene
EIF4E2
Gene Name
eukaryotic translation initiation factor 4E family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001276336.1 Intron NP_001263265.1
NM_001276337.1 Intron NP_001263266.1
NM_001282958.1 Intron NP_001269887.1
NM_004846.3 Intron NP_004837.1
XM_005246975.3 Intron XP_005247032.1
XM_011512206.2 Intron XP_011510508.1
XM_017005377.1 Intron XP_016860866.1
Gene
MIR5001
Gene Name
microRNA 5001
There are no transcripts associated with this gene.

Gene
TIGD1
Gene Name
tigger transposable element derived 1
There are no transcripts associated with this gene.

View Full Product Details