Product Details

SNP ID
rs37840
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:3312953 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCTCACTTAAAAAAATCATGTTCT[G/T]TTAATTCCTTTATCGCCTGGCCAAT
Phenotype
MIM: 612969 MIM: 601473
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TIGD7 PubMed Links

Gene Details

Gene
TIGD7
Gene Name
tigger transposable element derived 7
There are no transcripts associated with this gene.

Gene
ZNF75A
Gene Name
zinc finger protein 75a
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302109.1 Intron NP_001289038.1
NM_001324041.1 Intron NP_001310970.1
NM_001324085.1 Intron NP_001311014.1
NM_153028.3 Intron NP_694573.1
XM_011522646.2 Intron XP_011520948.1
XM_011522647.2 Intron XP_011520949.1
XM_017023642.1 Intron XP_016879131.1
XM_017023643.1 Intron XP_016879132.1
XM_017023644.1 Intron XP_016879133.1
XM_017023645.1 Intron XP_016879134.1

View Full Product Details