Product Details

SNP ID
rs29795
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.5:173150400 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TGGCCAGTTGGACCCCTGAGAAACA[A/G]TATGTCTGTGTCCTGTGTTTGCCTA
Phenotype
MIM: 603291
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BNIP1 PubMed Links

Gene Details

Gene
BNIP1
Gene Name
BCL2 interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001205.2 Intron NP_001196.2
NM_013978.2 Intron NP_053581.2
NM_013979.2 Intron NP_053582.2
NM_013980.2 Intron NP_053583.2
XM_011534638.1 Intron XP_011532940.1
XM_011534639.1 Intron XP_011532941.1

View Full Product Details