Product Details

SNP ID
rs272105
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:130846484 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGCACCCCCAGGCCAGCGCCCTC[C/A]CTCCAGGACCCTCAGGAACCTGCCT
Phenotype
MIM: 605216
Polymorphism
C/A, Transversion substitution
Allele Nomenclature
Literature Links
ARHGEF4 PubMed Links

Gene Details

Gene
ARHGEF4
Gene Name
Rho guanine nucleotide exchange factor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015320.3 Intron NP_056135.2
NM_032995.2 Intron NP_127462.1
XM_005263681.1 Intron XP_005263738.1
XM_005263685.3 Intron XP_005263742.1
XM_005263687.3 Intron XP_005263744.1
XM_005263688.2 Intron XP_005263745.1
XM_005263689.4 Intron XP_005263746.1
XM_011511274.2 Intron XP_011509576.1
XM_011511276.2 Intron XP_011509578.1
XM_011511277.1 Intron XP_011509579.1
XM_017004231.1 Intron XP_016859720.1
Gene
POTEI
Gene Name
POTE ankyrin domain family member I
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277406.1 Intron NP_001264335.1
XM_011511670.2 Intron XP_011509972.1
XM_017004732.1 Intron XP_016860221.1
XM_017004733.1 Intron XP_016860222.1
XM_017004734.1 Intron XP_016860223.1
XM_017004735.1 Intron XP_016860224.1

View Full Product Details