Product Details

SNP ID
rs220462
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.17:3724546 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GTTGCAGCCTCAGCCGTCCCGAGCG[A/G]CCTCCACCTCCCAGAAGTCTCCCTG
Phenotype
MIM: 609240 MIM: 604682
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GSG2 PubMed Links

Gene Details

Gene
GSG2
Gene Name
germ cell associated 2, haspin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031965.2 644 Missense Mutation GAC,GGC D204G NP_114171.2
Gene
ITGAE
Gene Name
integrin subunit alpha E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002208.4 644 Intron NP_002199.3
XM_011523823.2 644 Intron XP_011522125.1
XM_011523825.2 644 Intron XP_011522127.1
XM_011523827.2 644 Intron XP_011522129.1
XM_011523828.2 644 Intron XP_011522130.1
XM_017024586.1 644 Intron XP_016880075.1
XM_017024587.1 644 Intron XP_016880076.1

View Full Product Details