Product Details

SNP ID
rs416622
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:33005504 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGACCCCATCTCAGAAAAGACAGCC[C/T]CCCTGTTGCTGCCCCCTGCACTCCC
Phenotype
MIM: 142930
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HLA-DOA PubMed Links

Gene Details

Gene
HLA-DOA
Gene Name
major histocompatibility complex, class II, DO alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002119.3 2163 UTR 3 NP_002110.1

View Full Product Details