Product Details
- SNP ID
-
rs287986
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:9845477 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTGGTCTTAGGTACTTACACATAGG[C/T]TAAGGGATTTGTGAATTAGATTGAG
- Phenotype
-
MIM: 604904
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
TAF1B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79642340] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TAF1B
- Gene Name
- TATA-box binding protein associated factor, RNA polymerase I subunit B
View Full Product Details