Product Details

SNP ID
rs274866
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:48171470 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACATTTGAGGAGATGTCCTTAAGG[A/G]ACATCTGAGCTTAAGGGTCGTATTT
Phenotype
MIM: 126391
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C19orf68 PubMed Links

Gene Details

Gene
C19orf68
Gene Name
chromosome 19 open reading frame 68
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_199341.3 Intron NP_955373.3
XM_005259449.3 Intron XP_005259506.1
XM_006723204.3 Intron XP_006723267.1
XM_006723205.2 Intron XP_006723268.1
XM_011526936.2 Intron XP_011525238.1
XM_011526937.2 Intron XP_011525239.1
Gene
LIG1
Gene Name
DNA ligase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000234.2 Intron NP_000225.1
NM_001289063.1 Intron NP_001275992.1
NM_001289064.1 Intron NP_001275993.1
NM_001320970.1 Intron NP_001307899.1
NM_001320971.1 Intron NP_001307900.1
XM_017026809.1 Intron XP_016882298.1

View Full Product Details