Product Details

SNP ID
rs256539
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:189894094 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACAATTATCTCAGAAAATGTGTTT[A/G]CCAAATATATATCCTTGTACATTAG
Phenotype
MIM: 615117
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C2orf88 PubMed Links

Gene Details

Gene
C2orf88
Gene Name
chromosome 2 open reading frame 88
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042519.1 Intron NP_001035984.1
NM_001042520.1 Intron NP_001035985.1
NM_001042521.1 Intron NP_001035986.1
NM_032321.2 Intron NP_115697.2
XM_005246905.1 Intron XP_005246962.1
XM_006712795.2 Intron XP_006712858.1
XM_011511982.1 Intron XP_011510284.1
XM_011511983.1 Intron XP_011510285.1
XM_011511984.2 Intron XP_011510286.1
XM_011511985.2 Intron XP_011510287.1
XM_011511986.2 Intron XP_011510288.1
XM_017005096.1 Intron XP_016860585.1

View Full Product Details