Product Details

SNP ID
rs732288
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:67521915 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACCCCATGCCCCCCATGCCACCT[C/T]CCCTTCAGGGAACCAGTTCCTTCTC
Phenotype
MIM: 607314
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CABP2 PubMed Links

Gene Details

Gene
CABP2
Gene Name
calcium binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318496.1 Intron NP_001305425.1
NM_016366.2 Intron NP_057450.2

View Full Product Details