Product Details

SNP ID
rs743679
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:67585910 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTTTGCAATCATCTGGTGAGAGA[A/T]CCCAGCAAGGATGGACAGGCAGAAT
Phenotype
MIM: 134660
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
GSTP1 PubMed Links

Gene Details

Gene
GSTP1
Gene Name
glutathione S-transferase pi 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000852.3 Intron NP_000843.1

View Full Product Details