Product Details

SNP ID
rs492389
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65061408 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCCAACCAAGCCACTTTCAGATT[C/T]TTAACCTGTGGACACTGTGTGAGAT
Phenotype
MIM: 609374 MIM: 602640
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDCA5 PubMed Links

Gene Details

Gene
CDCA5
Gene Name
cell division cycle associated 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080668.3 Intron NP_542399.1
XM_005273733.3 Intron XP_005273790.1
XM_005273734.3 Intron XP_005273791.1
XM_011544743.2 Intron XP_011543045.1
XM_011544747.2 Intron XP_011543049.1
XM_011544748.2 Intron XP_011543050.1
XM_011544749.2 Intron XP_011543051.1
XM_017017158.1 Intron XP_016872647.1
Gene
NAALADL1
Gene Name
N-acetylated alpha-linked acidic dipeptidase-like 1
There are no transcripts associated with this gene.

View Full Product Details