Product Details

SNP ID
rs295505
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:139560431 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAGTTTTGTTTAAATTTTATGAAA[A/G]GCTGGTTAGGCAGGCAGACAATGGT
Phenotype
MIM: 608702
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC100507291 PubMed Links

Gene Details

Gene
LOC100507291
Gene Name
uncharacterized LOC100507291
There are no transcripts associated with this gene.

Gene
NMNAT3
Gene Name
nicotinamide nucleotide adenylyltransferase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001200047.2 1835 UTR 3 NP_001186976.1
NM_001320510.1 1835 UTR 3 NP_001307439.1
NM_001320511.1 1835 UTR 3 NP_001307440.1
NM_001320512.1 1835 UTR 3 NP_001307441.1
NM_001320513.1 1835 UTR 3 NP_001307442.1
NM_178177.4 1835 UTR 3 NP_835471.1
XM_011512790.1 1835 UTR 3 XP_011511092.1
XM_011512791.2 1835 UTR 3 XP_011511093.1
XM_017006328.1 1835 UTR 3 XP_016861817.1
XM_017006329.1 1835 UTR 3 XP_016861818.1
XM_017006330.1 1835 UTR 3 XP_016861819.1
XM_017006331.1 1835 UTR 3 XP_016861820.1
XM_017006332.1 1835 UTR 3 XP_016861821.1
XM_017006333.1 1835 UTR 3 XP_016861822.1
XM_017006334.1 1835 UTR 3 XP_016861823.1
XM_017006335.1 1835 UTR 3 XP_016861824.1
XM_017006336.1 1835 UTR 3 XP_016861825.1
XM_017006337.1 1835 UTR 3 XP_016861826.1
XM_017006338.1 1835 UTR 3 XP_016861827.1
XM_017006339.1 1835 UTR 3 XP_016861828.1
XM_017006340.1 1835 UTR 3 XP_016861829.1
XM_017006341.1 1835 UTR 3 XP_016861830.1
XM_017006342.1 1835 UTR 3 XP_016861831.1
XM_017006343.1 1835 UTR 3 XP_016861832.1
XM_017006344.1 1835 UTR 3 XP_016861833.1

View Full Product Details