Product Details

SNP ID
rs626216
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:93819163 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGACATTGGAGGCCTGAAAGGAGG[A/G]AGAAGAGAAAGGAATGTTCTAAGAC
Phenotype
MIM: 603810
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MED17 PubMed Links

Gene Details

Gene
MED17
Gene Name
mediator complex subunit 17
There are no transcripts associated with this gene.

Gene
VSTM5
Gene Name
V-set and transmembrane domain containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144871.1 Intron NP_001138343.1

View Full Product Details