Product Details

SNP ID
rs668920
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:63627804 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATACTTCTAGATATTTTTTATATT[G/T]GGGTAGGACTTTGATCCTTGCTAAG
Phenotype
MIM: 609369
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ATL3 PubMed Links

Gene Details

Gene
ATL3
Gene Name
atlastin GTPase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290048.1 3434 UTR 3 NP_001276977.1
NM_015459.4 3434 UTR 3 NP_056274.3
XM_006718493.1 3434 UTR 3 XP_006718556.1

View Full Product Details