Product Details
- SNP ID
-
rs638574
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:59950727 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GAGAGGTATGGGTGAAAATTGGTTG[A/G]GTCAGCTTTAAGGAAGTATGAGAAG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
OOSP1
PubMed Links
Gene Details
- Gene
- OOSP1
- Gene Name
- oocyte secreted protein 1, pseudogene
There are no transcripts associated with this gene.
View Full Product Details