Product Details

SNP ID
rs653502
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:68905909 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAACTGCCCTAGGGGGATGTAGGAG[A/G]AGCGGGATTGGGAGTGGAGCCAATT
Phenotype
MIM: 600502 MIM: 611834
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
IGHMBP2 PubMed Links

Gene Details

Gene
IGHMBP2
Gene Name
immunoglobulin mu binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002180.2 Intron NP_002171.2
XM_005273974.2 Intron XP_005274031.1
XM_005273975.3 Intron XP_005274032.1
XM_005273976.1 Intron XP_005274033.1
XM_011544994.1 Intron XP_011543296.1
XM_017017669.1 Intron XP_016873158.1
XM_017017670.1 Intron XP_016873159.1
XM_017017671.1 Intron XP_016873160.1
Gene
MRPL21
Gene Name
mitochondrial ribosomal protein L21
There are no transcripts associated with this gene.

View Full Product Details