Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_000993.4 | 3655 | Intron | NP_000984.1 | ||
NM_001098577.2 | 3655 | Intron | NP_001092047.1 | ||
NM_001099693.1 | 3655 | Intron | NP_001093163.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
XM_005263862.3 | 3655 | Missense Mutation | AGG,GGG | R1101G | XP_005263919.1 |
XM_005263863.4 | 3655 | Missense Mutation | AGG,GGG | R1094G | XP_005263920.1 |
XM_005263864.3 | 3655 | Missense Mutation | AGG,GGG | R1086G | XP_005263921.1 |
XM_011510507.1 | 3655 | Missense Mutation | AGG,GGG | R1073G | XP_011508809.1 |
XM_011510508.2 | 3655 | Intron | XP_011508810.1 | ||
XM_017003198.1 | 3655 | Missense Mutation | AGG,GGG | R1066G | XP_016858687.1 |
XM_017003199.1 | 3655 | Missense Mutation | AGG,GGG | R1051G | XP_016858688.1 |
XM_017003200.1 | 3655 | Intron | XP_016858689.1 | ||
XM_017003201.1 | 3655 | Intron | XP_016858690.1 | ||
XM_017003202.1 | 3655 | Intron | XP_016858691.1 |