Product Details

SNP ID
rs748503
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:71749708 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATTCCTTTGGGCCTAACTGGGGGG[C/T]TTAGAAGCAGGGCCATGTGTCCAGG
Phenotype
MIM: 615608 MIM: 605516
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C10orf54 PubMed Links

Gene Details

Gene
C10orf54
Gene Name
chromosome 10 open reading frame 54
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022153.1 2622 UTR 3 NP_071436.1
Gene
CDH23
Gene Name
cadherin-related 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171930.1 2622 Intron NP_001165401.1
NM_001171931.1 2622 Intron NP_001165402.1
NM_001171932.1 2622 Intron NP_001165403.1
NM_001171933.1 2622 Intron NP_001165404.1
NM_001171934.1 2622 Intron NP_001165405.1
NM_001171935.1 2622 Intron NP_001165406.1
NM_001171936.1 2622 Intron NP_001165407.1
NM_022124.5 2622 Intron NP_071407.4
NM_052836.3 2622 Intron NP_443068.1
XM_006717940.3 2622 Intron XP_006718003.1
XM_006717942.3 2622 Intron XP_006718005.1
XM_011540039.2 2622 Intron XP_011538341.1
XM_011540042.2 2622 Intron XP_011538344.1
XM_011540043.2 2622 Intron XP_011538345.1
XM_011540044.2 2622 Intron XP_011538346.1
XM_011540045.2 2622 Intron XP_011538347.1
XM_011540046.2 2622 Intron XP_011538348.1
XM_011540047.2 2622 Intron XP_011538349.1
XM_011540048.2 2622 Intron XP_011538350.1
XM_011540049.2 2622 Intron XP_011538351.1
XM_011540051.2 2622 Intron XP_011538353.1
XM_011540052.2 2622 Intron XP_011538354.1
XM_017016499.1 2622 Intron XP_016871988.1
XM_017016500.1 2622 Intron XP_016871989.1
XM_017016501.1 2622 Intron XP_016871990.1
XM_017016502.1 2622 Intron XP_016871991.1
XM_017016503.1 2622 Intron XP_016871992.1
XM_017016504.1 2622 Intron XP_016871993.1
XM_017016505.1 2622 Intron XP_016871994.1
XM_017016506.1 2622 Intron XP_016871995.1
XM_017016507.1 2622 Intron XP_016871996.1
XM_017016508.1 2622 Intron XP_016871997.1

View Full Product Details